Methyl 4-amino-2-(pyridin-3-ylamino)-1,3-thiazole-5-carboxylate
Lieferant: Apollo Scientific
OR111062-1G
APOSOR111062-1GEA
371
EUR
APOSOR111062-1G
Methyl 4-amino-2-(pyridin-3-ylamino)-1,3-thiazole-5-carboxylate
Methyl 4-amino-2-(pyridin-3-ylamino)-1,3-thiazole-5-carboxylate
Formel:
C₁₀H₁₀N₄O₂S Molecular Weight: 250,277 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28148237 CAS-Nummer: 1858251-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...