4-(4-Isopropylphenyl)-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific

OR111063-1G
APOSOR111063-1GEA 445 EUR
APOSOR111063-1G
4-(4-Isopropylphenyl)-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol
4-(4-Isopropylphenyl)-5-tetrahydro-2H-pyran-4-yl-4H-1,2,4-triazole-3-thiol
Formel: C₁₆H₂₁N₃OS
Molecular Weight: 303,422 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148238
CAS-Nummer: 1858251-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR