Ethyl 1-(5-methyl-6-phenylthieno[2,3-d]pyrimidin-4-yl)piperidine-4-carboxylate

Lieferant: Apollo Scientific

OR111064-1G
APOSOR111064-1GEA 346 EUR
APOSOR111064-1G
Ethyl 1-(5-methyl-6-phenylthieno[2,3-d]pyrimidin-4-yl)piperidine-4-carboxylate
Ethyl 1-(5-methyl-6-phenylthieno[2,3-d]pyrimidin-4-yl)piperidine-4-carboxylate
Formel: C₂₁H₂₃N₃O₂S
Molecular Weight: 381,491 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148252
CAS-Nummer: 1858257-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR