1-[4-Amino-2-(pyridin-3-ylamino)-1,3-thiazol-5-yl]-2,2-dimethylpropan-1-one
Lieferant: Apollo Scientific
OR111065-1G
APOSOR111065-1GEA
371
EUR
APOSOR111065-1G
1-[4-Amino-2-(pyridin-3-ylamino)-1,3-thiazol-5-yl]-2,2-dimethylpropan-1-one
1-[4-Amino-2-(pyridin-3-ylamino)-1,3-thiazol-5-yl]-2,2-dimethylpropan-1-one
Formel:
C₁₃H₁₆N₄OS Molecular Weight: 276,357 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28148239 CAS-Nummer: 1042734-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...