N-(5-Methyl-3-phenylisoxazol-4-yl)-N'-(tetrahydrofuran-2-ylmethyl)urea

Lieferant: Apollo Scientific

OR111066-1G
APOSOR111066-1GEA 445 EUR
APOSOR111066-1G
N-(5-Methyl-3-phenylisoxazol-4-yl)-N'-(tetrahydrofuran-2-ylmethyl)urea
N-(5-Methyl-3-phenylisoxazol-4-yl)-N'-(tetrahydrofuran-2-ylmethyl)urea
Formel: C₁₆H₁₉N₃O₃
Molecular Weight: 301,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148240
CAS-Nummer: 1858255-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR