1-(Oxolan-2-ylmethyl)-1H-pyrazol-4-amine

Lieferant: Apollo Scientific
Achtung

OR111069-1G
APOSOR111069-1GEA 371 EUR
APOSOR111069-1G
1-(Oxolan-2-ylmethyl)-1H-pyrazol-4-amine
1-(Oxolan-2-ylmethyl)-1H-pyrazol-4-amine
Formel: C₈H₁₃N₃O
Molecular Weight: 167,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11128191
CAS-Nummer: 1029413-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR