Methyl 4-amino-2-[(2-isopropyl-6-methylphenyl)amino]-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific

OR111070-1G
APOSOR111070-1GEA 310 EUR
APOSOR111070-1G
Methyl 4-amino-2-[(2-isopropyl-6-methylphenyl)amino]-1,3-thiazole-5-carboxylate
Methyl 4-amino-2-[(2-isopropyl-6-methylphenyl)amino]-1,3-thiazole-5-carboxylate
Formel: C₁₅H₁₉N₃O₂S
Molecular Weight: 305,395 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28148242
CAS-Nummer: 1858250-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR