Ethyl 1-(4-methoxybenzyl)piperidine-4-carboxylate
Lieferant: Apollo Scientific
OR111077-1G
APOSOR111077-1GEA
0
EUR
APOSOR111077-1G
Ethyl 1-(4-methoxybenzyl)piperidine-4-carboxylate
Ethyl 1-(4-methoxybenzyl)piperidine-4-carboxylate
Formel:
C₁₆H₂₃NO₃ Molecular Weight: 277,359 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00811156 CAS-Nummer: 148729-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...