Methyl 6-{[(4-chloro-3-nitrophenyl)carbamoyl]amino}nicotinate
Lieferant: Apollo Scientific
OR111080-1G
APOSOR111080-1GEA
0
EUR
APOSOR111080-1G
Methyl 6-{[(4-chloro-3-nitrophenyl)carbamoyl]amino}nicotinate
Methyl 6-{[(4-chloro-3-nitrophenyl)carbamoyl]amino}nicotinate
Formel:
C₁₄H₁₁ClN₄O₅ Molecular Weight: 350,714 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28347981 CAS-Nummer: 1858255-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...