N-(4-Chloro-3-nitrophenyl)-1H-imidazole-1-carboxamide
Lieferant: Apollo Scientific
OR111083-1G
OR111083-5G
OR111083-10G
APOSOR111083-1GEA
0
EUR
APOSOR111083-1G
APOSOR111083-5G
APOSOR111083-10G
N-(4-Chloro-3-nitrophenyl)-1H-imidazole-1-carboxamide
N-(4-Chloro-3-nitrophenyl)-1H-imidazole-1-carboxamide
Formel:
C₁₀H₇ClN₄O₃ Molecular Weight: 266.641 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23543204 CAS-Nummer: 1535430-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...