2-[4-(Oxan-4-yl)piperazin-1-yl]pyridin-3-amine

Lieferant: Apollo Scientific

OR111092-1G
APOSOR111092-1GEA 420 EUR
APOSOR111092-1G
2-[4-(Oxan-4-yl)piperazin-1-yl]pyridin-3-amine
2-[4-(Oxan-4-yl)piperazin-1-yl]pyridin-3-amine
Formel: C₁₄H₂₂N₄O
Molecular Weight: 262,351 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19059863
CAS-Nummer: 1486170-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR