4-(2,5-Dimethyl-4-nitro-pyrazol-3-yl)morpholine

Lieferant: Apollo Scientific

OR111099-1G
APOSOR111099-1GEA 0 EUR
APOSOR111099-1G
4-(2,5-Dimethyl-4-nitro-pyrazol-3-yl)morpholine
4-(2,5-Dimethyl-4-nitro-pyrazol-3-yl)morpholine
Formel: C₉H₁₄N₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00121959
CAS-Nummer: 312311-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR