6-(1H-Imidazol-1-yl)pyridin-3-amine

Lieferant: Apollo Scientific

OR111101-1G
APOSOR111101-1GEA 0 EUR
APOSOR111101-1G
6-(1H-Imidazol-1-yl)pyridin-3-amine
6-(1H-Imidazol-1-yl)pyridin-3-amine
Formel: C₈H₈N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08668590
CAS-Nummer: 681004-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR