Methyl 2-amino-4-chlorothieno[2,3-d]pyrimidine-6-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR111106-1G
APOSOR111106-1GEA
478
EUR
APOSOR111106-1G
Methyl 2-amino-4-chlorothieno[2,3-d]pyrimidine-6-carboxylate
Methyl 2-amino-4-chlorothieno[2,3-d]pyrimidine-6-carboxylate
Formel:
C₈H₆ClN₃O₂S Molecular Weight: 243,67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26395617 CAS-Nummer: 871231-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...