3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-thien-2-ylpropanoic acid
Lieferant: Apollo Scientific
OR111107-1G
APOSOR111107-1GEA
0
EUR
APOSOR111107-1G
3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-thien-2-ylpropanoic acid
3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-thien-2-ylpropanoic acid
Formel:
C₁₅H₁₃NO₃S Molecular Weight: 287,334 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02570954 CAS-Nummer: 478079-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...