3-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid

Lieferant: Apollo Scientific

OR111111-1G
APOSOR111111-1GEA 0 EUR
APOSOR111111-1G
3-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid
3-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid
Formel: C₁₆H₁₃NO₃
Molecular Weight: 267,279 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28954640
CAS-Nummer: 1858240-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR