4-(2-Methyl-1,3-thiazol-4-yl)benzene-1-sulphonamide
Lieferant: Apollo Scientific
OR111118-1G
APOSOR111118-1GEA
310
EUR
APOSOR111118-1G
4-(2-Methyl-1,3-thiazol-4-yl)benzene-1-sulphonamide
4-(2-Methyl-1,3-thiazol-4-yl)benzene-1-sulphonamide
Formel:
C₁₀H₁₀N₂O₂S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11650716 CAS-Nummer: 1094460-88-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...