3-Chlorothiophene-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR111119-1G
APOSOR111119-1GEA 310 EUR
APOSOR111119-1G
3-Chlorothiophene-2-carbohydrazide
3-Chlorothiophene-2-carbohydrazide
Formel: C₅H₅ClN₂OS
Molecular Weight: 176,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585402
CAS-Nummer: 478483-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR