5-Chloro-3-(morpholin-4-ylsulphonyl)pyridin-2-amine

Lieferant: Apollo Scientific

OR111127-1G
APOSOR111127-1GEA 0 EUR
APOSOR111127-1G
5-Chloro-3-(morpholin-4-ylsulphonyl)pyridin-2-amine
5-Chloro-3-(morpholin-4-ylsulphonyl)pyridin-2-amine
Formel: C₉H₁₂ClN₃O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD28954648
CAS-Nummer: 1858255-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR