N,N-Diisopropyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carboxamide

Lieferant: Apollo Scientific

OR111129-1G
APOSOR111129-1GEA 371 EUR
APOSOR111129-1G
N,N-Diisopropyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carboxamide
N,N-Diisopropyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carboxamide
Formel: C₁₅H₂₃NOS
Molecular Weight: 265,414 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29042494
CAS-Nummer: 1980039-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR