1-Pyridin-3-yl-1H-imidazole-2-thiol
Lieferant: Apollo Scientific
OR111141-1G
APOSOR111141-1GEA
0
EUR
APOSOR111141-1G
1-Pyridin-3-yl-1H-imidazole-2-thiol
1-Pyridin-3-yl-1H-imidazole-2-thiol
Formel:
C₈H₇N₃S Molecular Weight: 177,226 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00108600 CAS-Nummer: 17452-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...