1,5-Dimethyl-1H-pyrazole-3-carbohydrazide
Lieferant: Apollo Scientific
OR111143-1G
APOSOR111143-1GEA
0
EUR
APOSOR111143-1G
1,5-Dimethyl-1H-pyrazole-3-carbohydrazide
1,5-Dimethyl-1H-pyrazole-3-carbohydrazide
Formel:
C₆H₁₀N₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00221100 CAS-Nummer: 94447-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...