N-Methoxy-N-methyl-4-nitrobenzamide

Lieferant: Apollo Scientific

OR111155-5G
APOSOR111155-5GEA 0 EUR
APOSOR111155-5G
N-Methoxy-N-methyl-4-nitrobenzamide
N-Methoxy-N-methyl-4-nitrobenzamide
Formel: C₉H₁₀N₂O₄
Molecular Weight: 210,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07183934
CAS-Nummer: 52898-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR