N-(4-Nitrobenzoyl)morpholin
Lieferant: Apollo Scientific
Synonyme:
Morpholino(4-nitrophenyl)methanone
OR111156-5G
APOSOR111156-5GEA
0
EUR
APOSOR111156-5G
N-(4-Nitrobenzoyl)morpholin
N-(4-Nitrobenzoyl)morpholin
Formel:
C₁₁H₁₂N₂O₄ Molecular Weight: 236,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00451121 CAS-Nummer: 5397-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...