4-Imidazo[1,2-a]pyridin-2-ylbenzonitrile

Lieferant: Apollo Scientific
Achtung

OR111163-1G
APOSOR111163-1GEA 347 EUR
APOSOR111163-1G
4-Imidazo[1,2-a]pyridin-2-ylbenzonitrile
4-Imidazo[1,2-a]pyridin-2-ylbenzonitrile
Formel: C₁₄H₉N₃
Schmelzpunkt: 211…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05863434
CAS-Nummer: 55843-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR