Ethyl 4-imidazo[1,2-a]pyridin-2-ylbenzoate

Lieferant: Apollo Scientific

OR111165-1G
APOSOR111165-1GEA 0 EUR
APOSOR111165-1G
Ethyl 4-imidazo[1,2-a]pyridin-2-ylbenzoate
Ethyl 4-imidazo[1,2-a]pyridin-2-ylbenzoate
Formel: C₁₆H₁₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22573876
CAS-Nummer: 175153-33-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR