4-(6-Chloroimidazo[1,2-a]pyridin-2-yl)benzonitrile

Lieferant: Apollo Scientific

OR111168-10G OR111168-1G OR111168-5G
APOSOR111168-10GEA 0 EUR
APOSOR111168-10G APOSOR111168-1G APOSOR111168-5G
4-(6-Chloroimidazo[1,2-a]pyridin-2-yl)benzonitrile
4-(6-Chloroimidazo[1,2-a]pyridin-2-yl)benzonitrile
Formel: C₁₄H₈ClN₃
Molecular Weight: 253.686 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22043209
CAS-Nummer: 118000-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR