4-(6-Chloroimidazo[1,2-a]pyridin-2-yl)benzonitrile
Lieferant: Apollo Scientific
OR111168-10G
OR111168-1G
OR111168-5G
APOSOR111168-10GEA
0
EUR
APOSOR111168-10G
APOSOR111168-1G
APOSOR111168-5G
4-(6-Chloroimidazo[1,2-a]pyridin-2-yl)benzonitrile
4-(6-Chloroimidazo[1,2-a]pyridin-2-yl)benzonitrile
Formel:
C₁₄H₈ClN₃ Molecular Weight: 253.686 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22043209 CAS-Nummer: 118000-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...