N-Methyl-1H-indole-3-carboxamide

Lieferant: Apollo Scientific
Achtung

OR111177-1G
APOSOR111177-1GEA 337 EUR
APOSOR111177-1G
N-Methyl-1H-indole-3-carboxamide
N-Methyl-1H-indole-3-carboxamide
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174.199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12193321
CAS-Nummer: 85729-23-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR