7-Bromo-1-benzothiophene-2-carboxamide

Lieferant: Apollo Scientific
Achtung

OR111179-1G
APOSOR111179-1GEA 482 EUR
APOSOR111179-1G
7-Bromo-1-benzothiophene-2-carboxamide
7-Bromo-1-benzothiophene-2-carboxamide
Formel: C₉H₆BrNOS
Lagertemperatur: Raumtemperatur
MDL: MFCD29060036
CAS-Nummer: 1312904-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR