Luminol (3-Aminophthalhydrazid) 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: NSC 5064

OR11118-25G
APOSOR11118-25GEA 49.3 EUR
APOSOR11118-25G
Luminol (3-Aminophthalhydrazid) 98%
Luminol (3-Aminophthalhydrazid)
Formel: C₈H₇N₃O₂
Molecular Weight: 177.16 g/mol
Schmelzpunkt: 319…320 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006890
CAS-Nummer: 521-31-3
EINECS: 208-309-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR