7-Bromo-1-benzothiophene-2-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR111180-1G
APOSOR111180-1GEA
0
EUR
APOSOR111180-1G
7-Bromo-1-benzothiophene-2-carbaldehyde
7-Bromo-1-benzothiophene-2-carbaldehyde
Formel:
C₉H₅BrOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD17169310 CAS-Nummer: 19075-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...