N,N,1,2-Tetramethyl-1H-imidazole-4-sulphonamide

Lieferant: Apollo Scientific

OR111186-1G
APOSOR111186-1GEA 0 EUR
APOSOR111186-1G
N,N,1,2-Tetramethyl-1H-imidazole-4-sulphonamide
N,N,1,2-Tetramethyl-1H-imidazole-4-sulphonamide
Formel: C₇H₁₃N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09965509
CAS-Nummer: 936083-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR