3-Chloro-1H-indole-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloroindole-2-carboxylic acid

OR111187-1G
APOSOR111187-1GEA 128 EUR
APOSOR111187-1G
3-Chloro-1H-indole-2-carboxylic acid
3-Chloro-1H-indole-2-carboxylic acid
Formel: C₉H₆ClNO₂
Molecular Weight: 195,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11007738
CAS-Nummer: 28737-32-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR