Methyl 2-mercapto-1,3-benzoxazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR111188-1G
APOSOR111188-1GEA 192 EUR
APOSOR111188-1G
Methyl 2-mercapto-1,3-benzoxazole-5-carboxylate
Methyl 2-mercapto-1,3-benzoxazole-5-carboxylate
Formel: C₉H₇NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD07800461
CAS-Nummer: 72730-39-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR