5-Methoxybenzoxazole-2-thiol

Lieferant: Apollo Scientific

Synonyme: 5-Methoxybenzo[d]oxazole-2(3H)-thione

OR111189-1G
APOSOR111189-1GEA 209 EUR
APOSOR111189-1G
5-Methoxybenzoxazole-2-thiol
5-Methoxybenzoxazole-2-thiol
Formel: C₈H₇NO₂S
Molecular Weight: 181,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03030041
CAS-Nummer: 49559-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR