tert-Butyl 4-(5-amino-1,3,4-thiadiazol-2-yl)piperazine-1-carboxylate

Lieferant: Apollo Scientific

OR111193-1G OR111193-5G
APOSOR111193-1GEA 0 EUR
APOSOR111193-1G APOSOR111193-5G
tert-Butyl 4-(5-amino-1,3,4-thiadiazol-2-yl)piperazine-1-carboxylate
tert-Butyl 4-(5-amino-1,3,4-thiadiazol-2-yl)piperazine-1-carboxylate
Formel: C₁₁H₁₉N₅O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD24469692
CAS-Nummer: 244201-29-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR