tert-Butyl 4-(cyanoacetyl)piperazine-1-carboxylate

Lieferant: Apollo Scientific
Achtung

OR111203-1G
APOSOR111203-1GEA 75.4 EUR
APOSOR111203-1G
tert-Butyl 4-(cyanoacetyl)piperazine-1-carboxylate
tert-Butyl 4-(cyanoacetyl)piperazine-1-carboxylate
Formel: C₁₂H₁₉N₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20731084
CAS-Nummer: 159873-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR