5-Methoxy-N-methyl-1,3-benzoxazol-2-amine

Lieferant: Apollo Scientific
Achtung

OR111206-100MG
APOSOR111206-100MGEA 337 EUR
APOSOR111206-100MG
5-Methoxy-N-methyl-1,3-benzoxazol-2-amine
5-Methoxy-N-methyl-1,3-benzoxazol-2-amine
Formel: C₉H₁₀N₂O₂
Molecular Weight: 178,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19374710
CAS-Nummer: 1267987-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR