2,6-Dimethyl-2H-1,4-benzoxazin-3(4H)-one

Lieferant: Apollo Scientific

OR111207-1G
APOSOR111207-1GEA 0 EUR
APOSOR111207-1G
2,6-Dimethyl-2H-1,4-benzoxazin-3(4H)-one
2,6-Dimethyl-2H-1,4-benzoxazin-3(4H)-one
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01833988
CAS-Nummer: 17959-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR