(3,5-Dichloropyridin-2-yl)methanol

Lieferant: Apollo Scientific
Gefahr

OR111212-1G
APOSOR111212-1GEA 243 EUR
APOSOR111212-1G
(3,5-Dichloropyridin-2-yl)methanol
(3,5-Dichloropyridin-2-yl)methanol
Formel: C₆H₅Cl₂NO
Molecular Weight: 178.016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08448149
CAS-Nummer: 275383-87-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR