Ethyl-2-(6-bromoimidazo[1,2-a]pyridin-2-yl)acetate

Lieferant: Apollo Scientific

OR111216-1G
APOSOR111216-1GEA 0 EUR
APOSOR111216-1G
Ethyl-2-(6-bromoimidazo[1,2-a]pyridin-2-yl)acetate
Ethyl-2-(6-bromoimidazo[1,2-a]pyridin-2-yl)acetate
Formel: C₁₁H₁₁BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16172180
CAS-Nummer: 59128-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR