Ethyl imidazo[1,2-a]pyridin-2-ylacetate

Lieferant: Apollo Scientific
Achtung

OR111219-1G
APOSOR111219-1GEA 255 EUR
APOSOR111219-1G
Ethyl imidazo[1,2-a]pyridin-2-ylacetate
Ethyl imidazo[1,2-a]pyridin-2-ylacetate
Formel: C₁₁H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01113258
CAS-Nummer: 21755-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR