6-Piperidin-1-ylnicotinonitrile
Lieferant: Apollo Scientific
OR111228-1G
APOSOR111228-1GEA
0
EUR
APOSOR111228-1G
6-Piperidin-1-ylnicotinonitrile
6-Piperidin-1-ylnicotinonitrile
Formel:
C₁₁H₁₃N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09950318 CAS-Nummer: 501378-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...