Benzamidin Hydrochlorid Hydrat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Phenylamidin Hydrochlorid Hydrat, Benzoesäureamidin Hydrochlorid Hydrat, Benzamidiniumchlorid Hydrat
OR11123-25G
APOSOR11123-25GEA
29
EUR
APOSOR11123-25G
Benzamidin Hydrochlorid Hydrat
Benzamidin Hydrochlorid Hydrat
Formel:
C₆H₅C(=NH)NH₂·HCl·H₂O Molecular Weight: 174,63 g/mol Schmelzpunkt: 82…85 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00066285 CAS-Nummer: 206752-36-5 EINECS: 216-795-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...