4-Amino-N-pyridin-4-ylbenzenesulphonamide

Lieferant: Apollo Scientific

OR111230-1G
APOSOR111230-1GEA 0 EUR
APOSOR111230-1G
4-Amino-N-pyridin-4-ylbenzenesulphonamide
4-Amino-N-pyridin-4-ylbenzenesulphonamide
Formel: C₁₁H₁₁N₃O₂S
Molecular Weight: 249.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07365054
CAS-Nummer: 67638-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR