5-Azepan-1-yl-1,3,4-thiadiazol-2-amine

Lieferant: Apollo Scientific

OR111235-1G
APOSOR111235-1GEA 0 EUR
APOSOR111235-1G
5-Azepan-1-yl-1,3,4-thiadiazol-2-amine
5-Azepan-1-yl-1,3,4-thiadiazol-2-amine
Formel: C₈H₁₄N₄S
Molecular Weight: 198,289 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15732429
CAS-Nummer: 71125-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR