2-Azepan-1-ylnicotinonitrile

Lieferant: Apollo Scientific

OR111236-1G
APOSOR111236-1GEA 0 EUR
APOSOR111236-1G
2-Azepan-1-ylnicotinonitrile
2-Azepan-1-ylnicotinonitrile
Formel: C₁₂H₁₅N₃
Molecular Weight: 201,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08144877
CAS-Nummer: 684648-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR