2-[(4-Bromophenyl)methylene]malononitrile

Lieferant: Apollo Scientific

Synonyme: (4-Bromobenzylidene)malononitrile, 2-(4-Bromobenzylidene)malononitrile

OR111248-1G
APOSOR111248-1GEA 0 EUR
APOSOR111248-1G
2-[(4-Bromophenyl)methylene]malononitrile
2-[(4-Bromophenyl)methylene]malononitrile
Formel: C₁₀H₅BrN₂
Molecular Weight: 233,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00176213
CAS-Nummer: 2826-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR