(3-Bromobenzylidene)malononitrile

Lieferant: Apollo Scientific

OR111249-1G
APOSOR111249-1GEA 0 EUR
APOSOR111249-1G
(3-Bromobenzylidene)malononitrile
(3-Bromobenzylidene)malononitrile
Formel: C₁₀H₅BrN₂
Molecular Weight: 233,064 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00456782
CAS-Nummer: 2972-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR